Learn More
4-Iodobenzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1073018
Description
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.
Solubility
Partly miscible with water. Soluble in alcohol & ether.
Notes
It is sensitive to light. Incompatible with oxidizing agents and bases.
Specifications
4-Iodobenzoic acid | |
∼265°C (decomposition) | |
C7H5IO2 | |
50 g | |
Light sensitive | |
Partly miscible with water. Soluble in alcohol & ether. | |
OC(=O)C1=CC=C(I)C=C1 | |
248.02 | |
248.02 |
619-58-9 | |
2.18 | |
MFCD00002533 | |
1860232 | |
p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en | |
GHICCUXQJBDNRN-UHFFFAOYSA-N | |
4-iodobenzoic acid | |
12085 | |
97% |
Chemical Identifiers
619-58-9 | |
248.02 | |
GHICCUXQJBDNRN-UHFFFAOYSA-N | |
12085 | |
OC(=O)C1=CC=C(I)C=C1 |
C7H5IO2 | |
MFCD00002533 | |
p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en | |
4-iodobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-603-2
RTECSNumber : DH2976000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only