Learn More
4-Iodobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2441614
Description
It is used as intermediate in organic syntheses & in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as intermediate in organic syntheses & in medicine.
Solubility
Reacts with water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
Specifications
4-Iodobenzenesulfonyl chloride | |
79°C to 84°C | |
MFCD00007441 | |
UN3261 | |
Light and moisture sensitive | |
pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog | |
POXFXTSTVWDWIR-UHFFFAOYSA-N | |
4-iodobenzenesulfonyl chloride | |
7399 | |
97% |
98-61-3 | |
C6H4ClIO2S | |
25 g | |
2691661 | |
14,7486 | |
Reacts with water. | |
C1=CC(=CC=C1S(=O)(=O)Cl)I | |
302.51 | |
302.52 |
Chemical Identifiers
98-61-3 | |
302.51 | |
POXFXTSTVWDWIR-UHFFFAOYSA-N | |
7399 | |
C1=CC(=CC=C1S(=O)(=O)Cl)I |
C6H4ClIO2S | |
MFCD00007441 | |
pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog | |
4-iodobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-686-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only