missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodoanisole, 98+%

Manufacturer:  Alfa Aesar™ A1214414*

 View more versions of this product

Catalog No. AAA1214414



Description

Description

Applications
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

Solubility
Soluble in ethanol, ether, chloroform.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
Specifications

Specifications

4-Iodoanisole
≥98%
MFCD00001056
4-Methoxyiodobenzene
COC1=CC=C(C=C1)I
240°C
48°C to 52°C
234.04
25g
696-62-8
C7H7IO
1906692
SYSZENVIJHPFNL-UHFFFAOYSA-N
1-iodo-4-methoxybenzene
234.036
69676
>110°C (230°F)

Chemical Identifiers

696-62-8
234.036
SYSZENVIJHPFNL-UHFFFAOYSA-N
69676
COC1=CC=C(C=C1)I
C7H7IO
MFCD00001056
4-Methoxyiodobenzene
1-iodo-4-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents