missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodoaniline, TCI America™

$25.97 - $200.14

Chemical Identifiers

CAS 540-37-4
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007848
InChI Key VLVCDUSVTXIWGW-UHFFFAOYSA-N
Synonym p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
PubChem CID 10893
IUPAC Name 4-iodoaniline
SMILES C1=CC(=CC=C1N)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I004825G View Documents TCI America
I004825G
25g Each for $25.97
 
I0048250G View Documents TCI America
I0048250G
250g Each for $200.14
 
Specifications

Chemical Identifiers

540-37-4
219.025
VLVCDUSVTXIWGW-UHFFFAOYSA-N
10893
C1=CC(=CC=C1N)I
C6H6IN
MFCD00007848
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
4-iodoaniline

Specifications

Undesignated
25g
C6H6IN
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
C1=CC(=CC=C1N)I
219.025
219.03
≥99.0% (GC,T)
64°C
540-37-4
MFCD00007848
VLVCDUSVTXIWGW-UHFFFAOYSA-N
4-iodoaniline
10893
Crystal-Powder at 20°C
4-Iodoaniline
Safety and Handling

Safety and Handling

RTECSNumber : BY3850000

TSCA : Yes