missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodoaniline 99.0+%, TCI America™

Manufacturer:  Tci America I0048250G

 View more versions of this product

Catalog No. I0048250G



Specifications

Specifications

4-Iodoaniline
64°C
540-37-4
MFCD00007848
VLVCDUSVTXIWGW-UHFFFAOYSA-N
4-iodoaniline
10893
Crystal-Powder at 20°C
Undesignated
250g
C6H6IN
4-iodo-aniline, 4-iodo-benzenamine, 4-iodo-phenylamine, 4-iodobenzenamine, 4-iodophenylamine, aniline, 4-iodo, aniline, p-iodo, benzenamine, 4-iodo, p-aminophenyl iodide, p-iodoaniline
C1=CC(=CC=C1N)I
219.025
219.03
≥99.0% (GC,T)

Chemical Identifiers

540-37-4
219.025
VLVCDUSVTXIWGW-UHFFFAOYSA-N
10893
C1=CC(=CC=C1N)I
C6H6IN
MFCD00007848
4-iodo-aniline, 4-iodo-benzenamine, 4-iodo-phenylamine, 4-iodobenzenamine, 4-iodophenylamine, aniline, 4-iodo, aniline, p-iodo, benzenamine, 4-iodo, p-aminophenyl iodide, p-iodoaniline
4-iodoaniline
Safety and Handling

Safety and Handling

RTECSNumber : BY3850000

TSCA : Yes

Documents
Promotions

Promotions