missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Iodoacetophenone, 98%, ACROS Organics™

$34.07 - $34.07

Chemical Identifiers

CAS 13329-40-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H7IO
Linear Formula IC6H4C(O)CH3
Formula Weight 246.05
MDL Number MFCD00045320
Synonym 1-(4-Iodophenyl)ethanone
Chemical Name or Material 4'-Iodoacetophenone, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC278890050 SDS Thermo Fisher Scientific
AC278890050
Glass bottle 5g Each for $34.07
AC278890250 SDS Fisher Scientific Acros
278890250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13329-40-3
100.0
C8H7IO
246.05
1-(4-Iodophenyl)ethanone
≥97.5%
97.5
98%
IC6H4C(O)CH3
MFCD00045320
4'-Iodoacetophenone, 98%

Specifications

Crystalline Powder
5g
82° to 89°C
1-(4-Iodophenyl)ethanone
97.5
MFCD00045320
13329-40-3
IC6H4C(O)CH3
≥97.5%
246.05
Pale brown to brown
100.0
Glass bottle
Authentic
C8H7IO
98%
4'-Iodoacetophenone, 98%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 236-372-8

TSCA : TSCA