missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Iodoacetophenone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 278890250
Specifications
4'-Iodoacetophenone | |
Glass bottle | |
82.0°C to 89.0°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
IC6H4C(O)CH3 | |
1-4-iodo-phenyl-ethanone, 1-4-iodophenyl ethan-1-one, 1-4-iodophenyl ethanone, 1-acetyl-4-iodobenzene, 4'-iodoacetophenone, 4-iodo acetophenone, 4-iodoacetophenone, 4;-iodoacetophenone, ethanone, 1-4-iodophenyl, p-iodoacetophenone | |
CC(=O)C1=CC=C(C=C1)I | |
246.05 | |
246.05 | |
98% |
Authentic | |
Brown | |
25g | |
13329-40-3 | |
100.0 | |
C8H7IO | |
MFCD00045320 | |
JZJWCDQGIPQBAO-UHFFFAOYSA-N | |
1-(4-iodophenyl)ethanone | |
72869 | |
Crystalline Powder |
Chemical Identifiers
13329-40-3 | |
246.05 | |
JZJWCDQGIPQBAO-UHFFFAOYSA-N | |
72869 | |
CC(=O)C1=CC=C(C=C1)I |
C8H7IO | |
MFCD00045320 | |
1-4-iodo-phenyl-ethanone, 1-4-iodophenyl ethan-1-one, 1-4-iodophenyl ethanone, 1-acetyl-4-iodobenzene, 4'-iodoacetophenone, 4-iodo acetophenone, 4-iodoacetophenone, 4;-iodoacetophenone, ethanone, 1-4-iodophenyl, p-iodoacetophenone | |
1-(4-iodophenyl)ethanone |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 236-372-8
TSCA : TSCA