Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Iodoacetophenone, 98%

$39.03 - $411.35

Chemical Identifiers

Chemical Name or Material 4'-Iodoacetophenone
CAS 13329-40-3
Molecular Formula C8H7IO
Formula Weight 246.05
MDL Number MFCD00045320
Beilstein 1857412
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1679406

 
alfa aesar™
A1679406
5g Each for $39.03

AAA1679414

 
alfa aesar™
A1679414
25g Each for $128.82

AAA1679422

 
alfa aesar™
A1679422
100g Each for $411.35
Description & Specifications

Specifications

Formula Weight 246.05
Chemical Name or Material 4'-Iodoacetophenone
Assay 98%
Molecular Formula C8H7IO
MDL Number MFCD00045320
CAS 13329-40-3
Beilstein 1857412
Health Hazard 1 Warning
Melting Point 82° to 84°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.