Please login to your online account to display your discounted pricing

4-Iodo-m-xylene, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412500050

 View more versions of this product

Catalog No. AC412500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.06
Physical Form Liquid
Assay ≥94%
Quantity 5g
Color Clear yellow
TSCA TSCA
EINECS Number 224-141-4
CAS Min % 94.0
CAS Max % 100.0
Storage Information Product may darken on storage
Refractive Index 1.5970 to 1.5990 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00013706
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 95%
Name Note 95%
Molecular Formula C8H9I
CAS 4214-28-2
Chemical Name or Material 4-Iodo-m-xylene

Specifications

Chemical Name or Material 4-Iodo-m-xylene
Name Note 95%
CAS 4214-28-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H9I
Formula Weight 232.06
MDL Number MFCD00013706
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents