Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 4-Iodo-D-phenylalanine
CAS 62561-75-5
Formula Weight 291.09
MDL Number MFCD00063067
Synonym H-D-Phe(4-I)-OH; (R)-2-Amino-3-(4-iodophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6590703

 
alfa aesar™
H6590703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6590714

 
alfa aesar™
H6590714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6590706

 
alfa aesar™
H6590706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.09
Chemical Name or Material 4-Iodo-D-phenylalanine
CAS 62561-75-5
Physical Form Powder
Assay 95%
MDL Number MFCD00063067
Sensitivity Light Sensitive
Storage Note 1 2° to 8°C
Synonym H-D-Phe(4-I)-OH; (R)-2-Amino-3-(4-iodophenyl)propionic acid
Color Pale yellow
Melting Point 251°C
Quantity 1g
Safety and Handling

2° to 8°C