Please login to your online account to display your discounted pricing

4-Iodo-5-methyl-isoxazole, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC26311DE

 View more versions of this product

Catalog No. CC26311DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 208.98
Quantity 5g
Assay 97%
CAS Max % 100.0
Molecular Formula C4H4INO
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Iodo-5-methyl-isoxazole
MDL Number MFCD06411541
Synonym 4-Iodo-5-methyl-isoxazole (IUPAC)
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 97%
CAS 7064-38-2

Specifications

Chemical Name or Material 4-Iodo-5-methyl-isoxazole
CAS 7064-38-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H4INO
Formula Weight 208.98
MDL Number MFCD06411541
Synonym 4-Iodo-5-methyl-isoxazole (IUPAC)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.