missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-4',4''-dimethoxytriphenylamine 98.0+%, TCI America™

$277.05 - $277.05

Chemical Identifiers

CAS 201802-15-5
Molecular Formula C20H18INO2
Molecular Weight (g/mol) 431.273
InChI Key VGQDQJMYUYSINZ-UHFFFAOYSA-N
Synonym 4-Iodo-N,N-bis(4-methoxyphenyl)aniline
PubChem CID 44630018
IUPAC Name N-(4-iodophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07761G
View Documents Promotion Details
TCI America
I07761G
1 g
Each for $277.05
 
Description

Description

Specifications

Chemical Identifiers

201802-15-5
431.273
4-Iodo-N,N-bis(4-methoxyphenyl)aniline
N-(4-iodophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
C20H18INO2
VGQDQJMYUYSINZ-UHFFFAOYSA-N
44630018
COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)I

Specifications

201802-15-5
C20H18INO2
4-Iodo-N,N-bis(4-methoxyphenyl)aniline
COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)I
431.273
431.27
Crystalline Powder
111°C
1 g
VGQDQJMYUYSINZ-UHFFFAOYSA-N
N-(4-iodophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
44630018
≥98.0% (HPLC,N)
4-Iodo-4′,4′′-dimethoxytriphenylamine
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'