Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-3-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Iodo-3-(trifluoromethyl)aniline
CAS 155403-06-8
Formula Weight 287.02
MDL Number MFCD04972659
UN Number UN2811
Beilstein 11440140
Synonym 5-Amino-2-iodobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6442603

 
alfa aesar™
H6442603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6442606

 
alfa aesar™
H6442606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 287.02
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)aniline
Physical Form Solid
Assay 98%
MDL Number MFCD04972659
CAS 155403-06-8
UN Number UN2811
Beilstein 11440140
Color Off-white
Synonym 5-Amino-2-iodobenzotrifluoride
Quantity 1g
Sensitivity Light Sensitive
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and it is also an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Store in dark.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.