Please login to your online account to display your discounted pricing

4-Iodo-3-(trifluoromethyl)-1-H-pyrazole, 97%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC444820050

 View more versions of this product

Catalog No. AC444820050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 261.97
Quantity 5g
Molecular Formula C4H2F3IN2
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
CAS 866638-72-4
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Assay Percent Range 97%
Molecular Formula C4H2F3IN2
Formula Weight 261.97