Please login to your online account to display your discounted pricing

4-Iodo-3-(trifluoromethyl)-1-H-pyrazole, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Assay Percent Range 97%
Molecular Formula C4H2F3IN2
Formula Weight 261.97
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC444820050

  SDS
thermo fisher scientific
AC444820050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444820010

 
thermo fisher scientific
AC444820010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.97
Quantity 5g
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Assay Percent Range 97%
Molecular Formula C4H2F3IN2
CAS 866638-72-4