Please login to your online account to display your discounted pricing

4-Iodo-3-nitrophenol, 97%, ACROS Organics™

$476.57 - $476.57

Chemical Identifiers

Chemical Name or Material 4-Iodo-3-nitrophenol
CAS 113305-56-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4INO3
Formula Weight 265.01
MDL Number MFCD01320680
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC326280250

  SDS
thermo fisher scientific
AC326280250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC326280050

  SDS
thermo fisher scientific
326280050
Glass bottle 5g Each for $476.57
Description & Specifications

Specifications

Formula Weight 265.01
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 25g
Color Dark yellow to yellow-brown
Melting Point 153° to 159°C
Beilstein 06, II, 238
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Iodo-3-nitrophenol
Molecular Formula C6H4INO3
CAS 113305-56-9
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01320680
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.