missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-3-nitrophenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 326280050

 View more versions of this product

Catalog No. AC326280050


 

Specifications Description & Specifications

Specifications

4-Iodo-3-nitrophenol
96.0
Authentic
Crystalline Powder
97%
265.01
06, II, 238
153° to 159°C
113305-56-9
100.0
Glass bottle
≥96.0%
C6H4INO3
MFCD01320680
Dark yellow to yellow-brown
5g

Chemical Identifiers

113305-56-9
100.0
C6H4INO3
MFCD01320680
≥96.0%
96.0
97%
265.01
4-Iodo-3-nitrophenol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning