missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-3-nitrobenzoic acid, 97%

$242.34 - $985.81

Chemical Identifiers

CAS 35674-27-2
Molecular Formula C7H4INO4
Formula Weight 293.02
MDL Number MFCD00502399
Chemical Name or Material 4-Iodo-3-nitrobenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2562606 SDS Alfa Aesar™
B2562606
5g Each for $242.34
AAB2562614 SDS Alfa Aesar™
B2562614
25g Each for $985.81
Specifications

Description

Applications
4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.
Description & Specifications

Chemical Identifiers

35674-27-2
293.02
4-Iodo-3-nitrobenzoic acid
C7H4INO4
MFCD00502399

Specifications

35674-27-2
293.02
208° to 211°C
4-Iodo-3-nitrobenzoic acid
C7H4INO4
MFCD00502399
5g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning