Learn More
4-Iodo-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2562606
Description
4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from light, oxidizing agents.
Specifications
4-Iodo-3-nitrobenzoic acid | |
208°C to 211°C | |
MFCD00502399 | |
Light sensitive | |
Insoluble in water. | |
OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O | |
293.02 | |
293.02 |
35674-27-2 | |
C7H4INO4 | |
5 g | |
4-iodo-3-nitrobenzoic acid, 4-iodo-3-nitrobenzoicacid, 3-nitro-4-iodobenzoic acid, 4-iodo-3-nitro-benzoic acid, benzoic acid, 4-iodo-3-nitro, pubchem13471, acmc-209z8y, 4-iodo-3-nitro benzoic acid | |
DNMTZLCNLAIKQC-UHFFFAOYSA-N | |
4-iodo-3-nitrobenzoic acid | |
11208598 | |
97% |
Chemical Identifiers
35674-27-2 | |
293.02 | |
DNMTZLCNLAIKQC-UHFFFAOYSA-N | |
11208598 | |
OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O |
C7H4INO4 | |
MFCD00502399 | |
4-iodo-3-nitrobenzoic acid, 4-iodo-3-nitrobenzoicacid, 3-nitro-4-iodobenzoic acid, 4-iodo-3-nitro-benzoic acid, benzoic acid, 4-iodo-3-nitro, pubchem13471, acmc-209z8y, 4-iodo-3-nitro benzoic acid | |
4-iodo-3-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only