Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-3,5-dimethylisoxazole, 97%

$52.19 - $52.19

Chemical Identifiers

Chemical Name or Material 4-Iodo-3,5-dimethylisoxazole
CAS 10557-85-4
Molecular Formula C5H6INO
Formula Weight 223.01
MDL Number MFCD00173743
Synonym 3,5-Dimethyl-4-iodoisoxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1991103

 
alfa aesar™
A1991103
1g Each for $52.19
Description & Specifications

Specifications

Formula Weight 223.01
Chemical Name or Material 4-Iodo-3,5-dimethylisoxazole
Assay 97%
Molecular Formula C5H6INO
Melting Point 51° to 55°C
MDL Number MFCD00173743
CAS 10557-85-4
Quantity 1g
Synonym 3,5-Dimethyl-4-iodoisoxazole