Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-3,5-dimethyl-1H-pyrazole, 97%

$100.51 - $100.51

Chemical Identifiers

Chemical Name or Material 4-Iodo-3,5-dimethyl-1H-pyrazole
CAS 2033-45-6
Molecular Formula C5H7IN2
Formula Weight 222.04
MDL Number MFCD00040247
Synonym 3,5-Dimethyl-4-iodo-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2064788

 
alfa aesar™
B2064788
2.5g Each for $100.51
Description & Specifications

Specifications

Formula Weight 222.04
Chemical Name or Material 4-Iodo-3,5-dimethyl-1H-pyrazole
Assay 97%
Molecular Formula C5H7IN2
Melting Point 137° to 140°C
MDL Number MFCD00040247
CAS 2033-45-6
Quantity 2.5g
Odor Odorless
Synonym 3,5-Dimethyl-4-iodo-1H-pyrazole
RTECS Number UQ6517000