missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Iodo-3,5-bis-(trifluoromethyl)-1H-pyrazole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444790010
Specifications
4-Iodo-3, 5-bis-(trifluoromethyl)-1H-pyrazole | |
125°C to 126°C | |
1239511-06-8 | |
C5HF6IN2 | |
OTYORVJCWWESOF-UHFFFAOYSA-N | |
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole | |
45933690 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
3,5-bis trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3,5-bis trifluoromethyl-1h-pyrazole | |
C1(=C(NN=C1C(F)(F)F)C(F)(F)F)I | |
329.97 | |
329.97 |
Chemical Identifiers
1239511-06-8 | |
329.97 | |
3,5-bis trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3,5-bis trifluoromethyl-1h-pyrazole | |
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole |
C5HF6IN2 | |
OTYORVJCWWESOF-UHFFFAOYSA-N | |
45933690 | |
C1(=C(NN=C1C(F)(F)F)C(F)(F)F)I |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning