Please login to your online account to display your discounted pricing

4-Iodo-3,5-bis-(trifluoromethyl)-1H-pyrazole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Iodo-3, 5-bis-(trifluoromethyl)-1H-pyrazole
Assay Percent Range 97%
Molecular Formula C5HF6IN2
Formula Weight 329.97
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444790010

 
thermo fisher scientific
AC444790010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444790050

 
thermo fisher scientific
AC444790050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Iodo-3, 5-bis-(trifluoromethyl)-1H-pyrazole
CAS 1239511-06-8
Assay Percent Range 97%
Molecular Formula C5HF6IN2
Formula Weight 329.97
Quantity 1g