missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-3,5-bis-(trifluoromethyl)-1H-pyrazole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444790050

 View more versions of this product

Catalog No. AC444790050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Iodo-3, 5-bis-(trifluoromethyl)-1H-pyrazole
5g
96% min. (GC)
3,5-bis trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3,5-bis trifluoromethyl-1h-pyrazole
C1(=C(NN=C1C(F)(F)F)C(F)(F)F)I
329.97
329.97
125°C to 126°C
1239511-06-8
C5HF6IN2
OTYORVJCWWESOF-UHFFFAOYSA-N
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole
45933690
97%

Chemical Identifiers

1239511-06-8
329.97
3,5-bis trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3,5-bis trifluoromethyl-1h-pyrazole
4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole
C5HF6IN2
OTYORVJCWWESOF-UHFFFAOYSA-N
45933690
C1(=C(NN=C1C(F)(F)F)C(F)(F)F)I
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents