missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-2-(trifluoromethyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H55609MD

 View more versions of this product

Catalog No. AAH55609MD



Description

Description

Applications
4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-Iodo-2-(trifluoromethyl)benzonitrile
Light sensitive
250mg
C8H3F3IN
UN3439
Slightly soluble in water.
C1=CC(=C(C=C1I)C(F)(F)F)C#N
297.019
297.02
>110°C (230°F)
50°C to 55°C
101066-87-9
MFCD08458091
2-cyano-5-iodobenzotrifluoride
RBRTXKYFMXVIER-UHFFFAOYSA-N
4-iodo-2-(trifluoromethyl)benzonitrile
16218337
97%

Chemical Identifiers

101066-87-9
297.019
RBRTXKYFMXVIER-UHFFFAOYSA-N
16218337
C1=CC(=C(C=C1I)C(F)(F)F)C#N
C8H3F3IN
MFCD08458091
2-cyano-5-iodobenzotrifluoride
4-iodo-2-(trifluoromethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents