missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-2-methylaniline, 98+%

Manufacturer:  Alfa Aesar™ L0230606

 View more versions of this product

Catalog No. AAL0230606



Specifications

Specifications

4-Iodo-2-methylaniline
5g
C7H8IN
UN2811
2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
CC1=C(C=CC(=C1)I)N
233.052
233.05
84°C to 88°C
13194-68-8
MFCD00025299
2353618
BGKLFAQCHHCZRZ-UHFFFAOYSA-N
4-iodo-2-methylaniline
83221
≥98%

Chemical Identifiers

13194-68-8
233.052
BGKLFAQCHHCZRZ-UHFFFAOYSA-N
83221
CC1=C(C=CC(=C1)I)N
C7H8IN
MFCD00025299
2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
4-iodo-2-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents