Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-2-methoxypyridine-3-carboxaldehyde, 95%

Chemical Identifiers

Chemical Name or Material 4-Iodo-2-methoxypyridine-3-carboxaldehyde
CAS 158669-26-2
Formula Weight 263.03
MDL Number MFCD05662396
Beilstein 7020882
Synonym 4-Iodo-2-methoxynicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6698403

 
alfa aesar™
H6698403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66984MD

 
alfa aesar™
H66984MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.03
Chemical Name or Material 4-Iodo-2-methoxypyridine-3-carboxaldehyde
Physical Form Solid
Assay 95%
MDL Number MFCD05662396
CAS 158669-26-2
Beilstein 7020882
Synonym 4-Iodo-2-methoxynicotinaldehyde
Quantity 1g
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C