missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Iodo-2-methoxyaniline, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444800050
Specifications
4-Iodo-2-methoxyaniline | |
5g | |
96% min. (GC) | |
2-amino-5-iodoanisole, 2-amino-5-iodoanisole 4-iodo-2-methoxyaniline, 2-methoxy-4-iodoaniline, 4-iodo-2-methoxy-phenylamine, 4-iodo-o-anisidine, acmc-1aizp, benzenamine, 4-iodo-2-methoxy | |
COC1=C(C=CC(=C1)I)N | |
249.05 | |
249.05 |
48°C to 50°C | |
338454-80-1 | |
C7H8INO | |
AEPCMLLYVXZOLQ-UHFFFAOYSA-N | |
4-iodo-2-methoxyaniline | |
46737999 | |
97% |
Chemical Identifiers
338454-80-1 | |
249.05 | |
2-amino-5-iodoanisole, 2-amino-5-iodoanisole 4-iodo-2-methoxyaniline, 2-methoxy-4-iodoaniline, 4-iodo-2-methoxy-phenylamine, 4-iodo-o-anisidine, acmc-1aizp, benzenamine, 4-iodo-2-methoxy | |
4-iodo-2-methoxyaniline |
C7H8INO | |
AEPCMLLYVXZOLQ-UHFFFAOYSA-N | |
46737999 | |
COC1=C(C=CC(=C1)I)N |
Safety and Handling
GHS Signal Word: Warning