Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodo-2,6-dimethylphenol, 97%

$84.53 - $84.53

Chemical Identifiers

Chemical Name or Material 4-Iodo-2,6-dimethylphenol
CAS 10570-67-9
Molecular Formula C8H9IO
Formula Weight 248.06
MDL Number MFCD00094415
Synonym 4-Iodo-2,6-xylenol; 2,6-Dimethyl-4-iodophenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1984088

  SDS
alfa aesar™
A1984088
2.5g Each for $84.53
Description & Specifications

Specifications

Formula Weight 248.06
Quantity 2.5g
MDL Number MFCD00094415
Synonym 4-Iodo-2,6-xylenol; 2,6-Dimethyl-4-iodophenol
Health Hazard 1 Warning
CAS 10570-67-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 96° to 101°C
Assay 97%
Molecular Formula C8H9IO
Chemical Name or Material 4-Iodo-2,6-dimethylphenol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.