Learn More
4-Iodo-1H-pyrazole, 99%, Thermo Scientific Chemicals
$168.84 - $627.19
Chemical Identifiers
CAS | 3469-69-0 |
---|---|
Molecular Formula | C3H3IN2 |
Molecular Weight (g/mol) | 193.975 |
MDL Number | MFCD00005244 |
InChI Key | LLNQWPTUJJYTTE-UHFFFAOYSA-N |
Synonym | 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 |
PubChem CID | 77022 |
ChEBI | CHEBI:45260 |
IUPAC Name | 4-iodo-1H-pyrazole |
SMILES | C1=C(C=NN1)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5636906
|
Thermo Scientific Chemicals
H5636906 |
5 g |
Each for $168.84
|
|
|||||
AAH5636914
|
Thermo Scientific Chemicals
H5636914 |
25 g |
Each for $627.19
|
|
|||||
Description
4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.
Solubility
Soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
Chemical Identifiers
3469-69-0 | |
193.975 | |
LLNQWPTUJJYTTE-UHFFFAOYSA-N | |
77022 | |
4-iodo-1H-pyrazole |
C3H3IN2 | |
MFCD00005244 | |
4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 | |
CHEBI:45260 | |
C1=C(C=NN1)I |
Specifications
3469-69-0 | |
C3H3IN2 | |
5 g | |
4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295 | |
LLNQWPTUJJYTTE-UHFFFAOYSA-N | |
4-iodo-1H-pyrazole | |
77022 | |
193.97 | |
4-Iodo-1H-pyrazole |
108°C to 110°C | |
MFCD00005244 | |
Light sensitive | |
Soluble in water. | |
C1=C(C=NN1)I | |
193.975 | |
CHEBI:45260 | |
99% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 222-434-1
RTECSNumber : UQ7145000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only