Please login to your online account to display your discounted pricing

4-Iodo-1,3,5-trimethyl-1H-pyrazole, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC14711DA

 View more versions of this product

Catalog No. CC14711DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 236.05
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C6H9IN2
CAS 51660-65-2
CAS Min % 96.5
MDL Number MFCD00626320
Packaging Amber Glass Bottle
Chemical Name or Material 4-Iodo-1, 3, 5-trimethyl-1H-pyrazole
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-Iodo-1, 3, 5-trimethyl-1H-pyrazole
CAS 51660-65-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9IN2
Formula Weight 236.05
MDL Number MFCD00626320