missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-1,3,5-trimethyl-1H-pyrazole, 97%, Maybridge

Chemical Identifiers

CAS 51660-65-2
Molecular Formula C6H9IN2
Molecular Weight (g/mol) 236.056
MDL Number MFCD00626320
InChI Key RUOGLHMRBURIEQ-UHFFFAOYSA-N
Synonym 4-iodo-1,3,5-trimethyl-1h-pyrazole, 1,3,5-trimethyl-4-iodo-1h-pyrazole, amth061, 4-iodanyl-1,3,5-trimethyl-pyrazole, 1h-pyrazole,4-iodo-1,3,5-trimethyl, 1h-pyrazole, 4-iodo-1,3,5-trimethyl
PubChem CID 616702
IUPAC Name 4-iodo-1,3,5-trimethylpyrazole
SMILES CC1=C(C(=NN1C)C)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14711EA SDS View Documents Fisher Scientific Company
CC14711EA
10g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

51660-65-2
236.056
RUOGLHMRBURIEQ-UHFFFAOYSA-N
616702
CC1=C(C(=NN1C)C)I
C6H9IN2
MFCD00626320
4-iodo-1,3,5-trimethyl-1h-pyrazole, 1,3,5-trimethyl-4-iodo-1h-pyrazole, amth061, 4-iodanyl-1,3,5-trimethyl-pyrazole, 1h-pyrazole,4-iodo-1,3,5-trimethyl, 1h-pyrazole, 4-iodo-1,3,5-trimethyl
4-iodo-1,3,5-trimethylpyrazole

Specifications

Amber Glass Bottle
51660-65-2
100.0
C6H9IN2
4-iodo-1,3,5-trimethyl-1h-pyrazole, 1,3,5-trimethyl-4-iodo-1h-pyrazole, amth061, 4-iodanyl-1,3,5-trimethyl-pyrazole, 1h-pyrazole,4-iodo-1,3,5-trimethyl, 1h-pyrazole, 4-iodo-1,3,5-trimethyl
CC1=C(C(=NN1C)C)I
236.056
236.05
4-Iodo-1, 3, 5-trimethyl-1H-pyrazole
10g
96.5
97%
MFCD00626320
RUOGLHMRBURIEQ-UHFFFAOYSA-N
4-iodo-1,3,5-trimethylpyrazole
616702
97%