missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Iodo-1,3,5-trimethyl-1H-pyrazole, 97%, Maybridge™
Manufacturer: Fisher Scientific Company CC14711EA
Specifications
4-Iodo-1, 3, 5-trimethyl-1H-pyrazole | |
10g | |
96.5 | |
97% | |
MFCD00626320 | |
RUOGLHMRBURIEQ-UHFFFAOYSA-N | |
4-iodo-1,3,5-trimethylpyrazole | |
616702 | |
97% |
Amber Glass Bottle | |
51660-65-2 | |
100.0 | |
C6H9IN2 | |
1,3,5-trimethyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1,3,5-trimethyl, 1h-pyrazole,4-iodo-1,3,5-trimethyl, 4-iodanyl-1,3,5-trimethyl-pyrazole, 4-iodo-1,3,5-trimethyl-1h-pyrazole, amth061 | |
CC1=C(C(=NN1C)C)I | |
236.056 | |
236.05 |
Chemical Identifiers
51660-65-2 | |
236.056 | |
RUOGLHMRBURIEQ-UHFFFAOYSA-N | |
616702 | |
CC1=C(C(=NN1C)C)I |
C6H9IN2 | |
MFCD00626320 | |
1,3,5-trimethyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1,3,5-trimethyl, 1h-pyrazole,4-iodo-1,3,5-trimethyl, 4-iodanyl-1,3,5-trimethyl-pyrazole, 4-iodo-1,3,5-trimethyl-1h-pyrazole, amth061 | |
4-iodo-1,3,5-trimethylpyrazole |