Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Imidazolyl)acetonitrile, 97%

Chemical Identifiers

Chemical Name or Material (4-Imidazolyl)acetonitrile
CAS 18502-05-1
Molecular Formula C5H5N3
Formula Weight 107.11
MDL Number MFCD00070306
UN Number UN3439
Synonym 4-(Cyanomethyl)imidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3299206

  SDS
alfa aesar™
H3299206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3299203

  SDS
alfa aesar™
H3299203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 107.11
Quantity 5g
Odor Odorless
Molecular Formula C5H5N3
Chemical Name or Material (4-Imidazolyl)acetonitrile
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 18502-05-1
Synonym 4-(Cyanomethyl)imidazole
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Assay 97%
MDL Number MFCD00070306
Melting Point 136° to 138°C
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN3439

Applications
(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.