Please login to your online account to display your discounted pricing

4'-(Imidazol-1-yl)acetophenone, 97%, ACROS Organics™

$96.72 - $96.72

Chemical Identifiers

Chemical Name or Material 4'-(Imidazol-1-yl)acetophenone
Name Note 97%
CAS 10041-06-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H10N2O
Formula Weight 186.21
MDL Number MFCD00005282
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179800050

  SDS
thermo fisher scientific
179800050
Glass bottle 5g Each for $96.72
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 186.21
Quantity 5g
Color White to yellow
EINECS Number 233-123-5
TSCA TSCA
Melting Point 112° to 119°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4'-(Imidazol-1-yl)acetophenone
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O
MDL Number MFCD00005282
CAS 10041-06-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.