Please login to your online account to display your discounted pricing

4-IBP, Tocris

σ ligand, σ1 > σ2

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material 4-IBP
Assay Percent Range >99%
Molecular Formula C19H21IN2O
Formula Weight 420.29
Synonym N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

074810

 
r & d systems
RDS074810
10mg Each for $149.90

074850

 
r & d systems
RDS074850
50mg Each for $632.23
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 420.29
Molecular Formula C19H21IN2O
CAS 155798-08-6
Synonym N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
Recommended Storage Desiccate at +4°C
Chemical Name or Material 4-IBP
Assay Percent Range >99%
Safety and Handling

Desiccate at +4°C