Please login to your online account to display your discounted pricing

4-IBP, Tocris


σ ligand, σ1 > σ2

Quantity 50mg

Manufacturer: r & d systems  RDS074850

 View more versions of this product

Catalog No. 074850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 420.29
Chemical Name or Material 4-IBP
Assay Percent Range >99%
Molecular Formula C19H21IN2O
CAS 155798-08-6
Synonym N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 4-IBP
Assay Percent Range >99%
Molecular Formula C19H21IN2O
Formula Weight 420.29
Synonym N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
Accessories

Desiccate at +4°C