Please login to your online account to display your discounted pricing

4'-Hydroxypropiophenone, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4';-Hydroxypropiophenone
Name Note 98+%
CAS 70-70-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H10O2
Linear Formula HOC6H4COC2H5
Formula Weight 150.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121815000

  SDS
thermo fisher scientific
121815000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 150.18
Quantity 500g
Color White
TSCA TSCA
RTECS Number UH1925000
EINECS Number 200-743-2
Melting Point 147° to 151°C
Beilstein 08, 102
Merck Index 15, 7149
CAS Min % 98.0
CAS Max % 100.0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Plastic bottle
Infrared Spectrum Authentic
Synonym Paroxypropione
Name Note 98+%
CAS 70-70-2
Assay Percent Range 98+%
MDL Number MFCD00002361
Linear Formula HOC6H4COC2H5
Chemical Name or Material 4';-Hydroxypropiophenone
Molecular Formula C9H10O2