Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxypiperidine, 97%

Chemical Identifiers

Chemical Name or Material 4-Hydroxypiperidine
CAS 5382-16-1
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD00005999
UN Number UN3263
Beilstein 102738
Synonym 4-Piperidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2272306

  SDS
alfa aesar™
B2272306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2272314

  SDS
alfa aesar™
B2272314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2272322

  SDS
alfa aesar™
B2272322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.15
Beilstein 102738
Molecular Formula C5H11NO
MDL Number MFCD00005999
Boiling Point 108° to 114°C (10mmHg)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flash Point 107°C (224°F)
Health Hazard 1 Danger
Melting Point 86° to 90°C
Quantity 5g
pH 11.5 to 11.7
UN Number UN3263
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-Piperidinol
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
CAS 5382-16-1
Odor Amine-like
Assay 97%
Chemical Name or Material 4-Hydroxypiperidine
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.