missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyphthalonitrile, TCI America™

$122.22 - $317.78

Chemical Identifiers

CAS 30757-50-7
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
MDL Number MFCD00191653
InChI Key FTVOPKROFUTOKY-UHFFFAOYSA-N
Synonym 3,4-Dicyanophenol
PubChem CID 3875765
IUPAC Name 4-hydroxybenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1O)C#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08131G View Documents TCI America
H08131G
1g Each for $122.22
 
H08135G View Documents TCI America
H08135G
5g Each for $317.78
 
Specifications

Chemical Identifiers

30757-50-7
144.133
FTVOPKROFUTOKY-UHFFFAOYSA-N
3875765
C1=CC(=C(C=C1O)C#N)C#N
C8H4N2O
MFCD00191653
3,4-Dicyanophenol
4-hydroxybenzene-1,2-dicarbonitrile

Specifications

Undesignated
1g
C8H4N2O
3,4-Dicyanophenol
C1=CC(=C(C=C1O)C#N)C#N
144.133
144.13
≥97.0% (GC)
212°C
30757-50-7
MFCD00191653
FTVOPKROFUTOKY-UHFFFAOYSA-N
4-hydroxybenzene-1,2-dicarbonitrile
3875765
Crystal-Powder at 20°C
4-Hydroxyphthalonitrile
Safety and Handling

Safety and Handling

TSCA : No