Learn More
4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 412130050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Hydroxyphthalic acid | |
97.5 | |
191.0°C to 194.0°C | |
Authentic | |
Glass bottle | |
5 g | |
Solubility in water: insoluble. | |
C1=CC(=C(C=C1O)C(=O)O)C(=O)O | |
182.13 | |
CHEBI:27600 | |
98% |
610-35-5 | |
100.0 | |
White | |
98% | |
C8H6O5 | |
1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy | |
MWRVRCAFWBBXTL-UHFFFAOYSA-N | |
4-hydroxyphthalic acid | |
11881 | |
182.13 | |
Powder |
Chemical Identifiers
610-35-5 | |
182.13 | |
1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy | |
CHEBI:27600 | |
C1=CC(=C(C=C1O)C(=O)O)C(=O)O |
C8H6O5 | |
MWRVRCAFWBBXTL-UHFFFAOYSA-N | |
11881 | |
4-hydroxyphthalic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 210-221-6
RUO – Research Use Only