Learn More
4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1509122
Description
4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
4-Hydroxyphthalic acid | |
∼190°C (decomposition) | |
MFCD00013984 | |
1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy | |
MWRVRCAFWBBXTL-UHFFFAOYSA-N | |
4-hydroxyphthalic acid | |
11881 | |
182.13 |
610-35-5 | |
C8H6O5 | |
100 g | |
Slightly miscible with water. | |
C1=CC(=C(C=C1O)C(=O)O)C(=O)O | |
182.131 | |
CHEBI:27600 | |
98% |
Chemical Identifiers
610-35-5 | |
182.131 | |
MWRVRCAFWBBXTL-UHFFFAOYSA-N | |
11881 | |
4-hydroxyphthalic acid |
C8H6O5 | |
MFCD00013984 | |
1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy | |
CHEBI:27600 | |
C1=CC(=C(C=C1O)C(=O)O)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 210-221-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only