Please login to your online account to display your discounted pricing

4-Hydroxyphenylpyruvic Acid 98%, ACROS Organics™

$308.87 - $308.87

Chemical Identifiers

Chemical Name or Material 4-Hydroxyphenylpyruvic acid
Name Note 98%
CAS 156-39-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8O4
Linear Formula HOC6H4CH2COCO2H
Formula Weight 180.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151300010

  SDS
fisher scientific acros
151300010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151300050

  SDS
thermo fisher scientific
151300050
Glass bottle 5g Each for $308.87
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 180.16
Quantity 1g
Color Gray to orange-brown
EINECS Number 205-852-9
RTECS Number UZ1000000
Beilstein 10, 955
CAS Max % 100.0
Chemical Name or Material 4-Hydroxyphenylpyruvic acid
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H8O4
Linear Formula HOC6H4CH2COCO2H
MDL Number MFCD00002591
CAS 156-39-8
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.