missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyphenyldimethylsulfonium Methyl Sulfate, TCI America™

$184.86 - $554.59

Chemical Identifiers

CAS 32279-04-2
Molecular Formula C9H14O5S2
Molecular Weight (g/mol) 266.326
InChI Key PKQZVASULXKBJV-UHFFFAOYSA-N
Synonym Dimethyl-4-hydroxyphenylsulfonium Methyl Sulfate
PubChem CID 21915988
IUPAC Name (4-hydroxyphenyl)-dimethylsulfanium;methyl sulfate
SMILES COS(=O)(=O)[O-].C[S+](C)C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H102225G View Documents TCI America
H102225G
25g Each for $554.59
 
H10225G View Documents TCI America
H10225G
5g Each for $184.86
 
Specifications

Chemical Identifiers

32279-04-2
266.326
Dimethyl-4-hydroxyphenylsulfonium Methyl Sulfate
(4-hydroxyphenyl)-dimethylsulfanium;methyl sulfate
C9H14O5S2
PKQZVASULXKBJV-UHFFFAOYSA-N
21915988
COS(=O)(=O)[O-].C[S+](C)C1=CC=C(C=C1)O

Specifications

White-Yellow
25g
C9H14O5S2
PKQZVASULXKBJV-UHFFFAOYSA-N
(4-hydroxyphenyl)-dimethylsulfanium;methyl sulfate
21915988
Crystal-Powder at 20°C
4-Hydroxyphenyldimethylsulfonium Methyl Sulfate
92°C
32279-04-2
Dimethyl-4-hydroxyphenylsulfonium Methyl Sulfate
COS(=O)(=O)[O-].C[S+](C)C1=CC=C(C=C1)O
266.326
266.33
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No