Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxyphenylacetone, 98+%

$509.50 - $509.50

Chemical Identifiers

Chemical Name or Material 4-Hydroxyphenylacetone
CAS 770-39-8
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00210439
Synonym 1-(4-Hydroxyphenyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1893614

  SDS
alfa aesar™
A1893614
25g Each for $509.50
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material 4-Hydroxyphenylacetone
Molecular Formula C9H10O2
MDL Number MFCD00210439
CAS 770-39-8
Synonym 1-(4-Hydroxyphenyl)-2-propanone
Quantity 25g
Assay ≥98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.