Please login to your online account to display your discounted pricing

4-Hydroxyphenylacetamide, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  254272500

 View more versions of this product

Catalog No. AC254272500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 151.16
Quantity 250g
Color Beige
Beilstein 10, III, 434
EINECS Number 241-235-0
TSCA TSCA
RTECS Number AC3851500
Melting Point 175° to 178°C
CAS Max % 100.0
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 4-Hydroxybenzeneacetamide
Name Note 99%
CAS 17194-82-0
Assay Percent Range 99%
MDL Number MFCD00017145
Molecular Formula C8H9NO2
Chemical Name or Material 4-Hydroxyphenylacetamide

Specifications

Chemical Name or Material 4-Hydroxyphenylacetamide
Name Note 99%
CAS 17194-82-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9NO2
Formula Weight 151.16
MDL Number MFCD00017145
Synonym 4-Hydroxybenzeneacetamide