missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyphenylacetamide, 99%, ACROS Organics™

$22.16 - $22.16

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00017145
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxybenzeneacetamide, 4-hydroxyphenyl acetamide, 4-hydroxyphenylacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC254270050
SDS View Documents
Thermo Fisher Scientific
254270050
5g Glass bottle
Each for $22.16
 
Description

Description

Specifications

Chemical Identifiers

17194-82-0
151.16
YBPAYPRLUDCSEY-UHFFFAOYSA-N
86986
C1=CC(=CC=C1CC(=O)N)O
C8H9NO2
MFCD00017145
2-4-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxybenzeneacetamide, 4-hydroxyphenyl acetamide, 4-hydroxyphenylacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, unii-0hy0n4itn4
2-(4-hydroxyphenyl)acetamide

Specifications

Authentic
Beige
5g
98.5
98.5% min. (HPLC)
MFCD00017145
2-4-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxybenzeneacetamide, 4-hydroxyphenyl acetamide, 4-hydroxyphenylacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, unii-0hy0n4itn4
YBPAYPRLUDCSEY-UHFFFAOYSA-N
2-(4-hydroxyphenyl)acetamide
86986
Fine Crystalline Powder
4-Hydroxyphenylacetamide, 99%
Glass bottle
176.0°C to 180.0°C
17194-82-0
100.0
C8H9NO2
10, III, 434
Solubility in water: soluble. Other solubilities: soluble in ethanol,methanol
C1=CC(=CC=C1CC(=O)N)O
151.16
151.16
99%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 241-235-0

RTECSNumber : AC3851500

TSCA : TSCA

SDS