missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxyphenylacetamide, 99%

Manufacturer:  Alfa Aesar™ A1526630

 View more versions of this product

Catalog No. AAA1526630



Specifications

Specifications

4-Hydroxyphenylacetamide
250g
C8H9NO2
2085984
YBPAYPRLUDCSEY-UHFFFAOYSA-N
2-(4-hydroxyphenyl)acetamide
86986
99%
173°C to 175°C
17194-82-0
MFCD00017145
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
C1=CC(=CC=C1CC(=O)N)O
151.165
151.17

Chemical Identifiers

17194-82-0
151.165
YBPAYPRLUDCSEY-UHFFFAOYSA-N
86986
C1=CC(=CC=C1CC(=O)N)O
C8H9NO2
MFCD00017145
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
2-(4-hydroxyphenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AC3851500

TSCA : TSCA

Documents