missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyphenylacetamide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1526618

 View more versions of this product

Catalog No. AAA1526618



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Hydroxyphenylacetamide
173°C to 175°C
MFCD00017145
2085984
YBPAYPRLUDCSEY-UHFFFAOYSA-N
2-(4-hydroxyphenyl)acetamide
86986
99%
17194-82-0
C8H9NO2
50 g
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
C1=CC(=CC=C1CC(=O)N)O
151.165
151.17

Chemical Identifiers

17194-82-0
151.165
YBPAYPRLUDCSEY-UHFFFAOYSA-N
86986
C1=CC(=CC=C1CC(=O)N)O
C8H9NO2
MFCD00017145
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
2-(4-hydroxyphenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-235-0

RTECSNumber : AC3851500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only