Learn More
4-Hydroxyphenethyl alcohol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 182500050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Hydroxyphenethyl alcohol | |
501-94-0 | |
100.0 | |
White to Beige | |
97.5% min. (GC) | |
C8H10O2 | |
MFCD00002902 | |
06, IV, 5936 | |
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol | |
YCCILVSKPBXVIP-UHFFFAOYSA-N | |
4-(2-hydroxyethyl)phenol | |
10393 | |
138.17 | |
Crystalline Powder or Crystals |
98% | |
97.5 | |
89.0°C to 92.0°C | |
Authentic | |
Glass bottle | |
HOC6H4CH2CH2OH | |
5 g | |
15, 10022 | |
Solubility in water: 124g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,chloroform,,diethyl ether,petroleum ether | |
C1=CC(=CC=C1CCO)O | |
138.17 | |
CHEBI:1879 | |
98% |
Chemical Identifiers
501-94-0 | |
138.17 | |
YCCILVSKPBXVIP-UHFFFAOYSA-N | |
10393 | |
4-(2-hydroxyethyl)phenol |
C8H10O2 | |
MFCD00002902 | |
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol | |
CHEBI:1879 | |
C1=CC(=CC=C1CCO)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
EINECSNumber : 207-930-8
TSCA : TSCA
RUO – Research Use Only